Learn More
Buffer solution pH 7.00 (+/-0.024 @ 25oC) Colored Green Specpure NIST Traceable
Description
Buffer solutions play a substantial role in industrial and physiological processes. Buffer solutions of known and precise pH value allow to set the testing system to measure the samples of unknown pH more precisely and accurately.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Chemical Name or Material | Buffer solution |
Name Note | NIST Traceable |
CAS | 1336-21-6 |
Color | Green |
Physical Form | Liquid |
Quantity | 500 mL |
Molecular Formula | H5NO |
MDL Number | MFCD00066650 |
Traceability to NIST | Yes |
Vapor Pressure | 23 hPa (17mm Hg) at 20°C |
Show More |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.