Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buffer solution pH 9.2, Borate buffer, traceable to NIST, ready to use

Catalog No. p-3598324
Click to view available options
Quantity:
500 mL
1 L

CAS: 1330-43-4

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Buffer solution pH 9.2
Name Note Borate buffer, traceable to NIST, ready to use
CAS 1330-43-4, 7732-18-5, 7647-14-5
Density 1.0000g/mL
Quantity 500 mL
MDL Number MFCD00163147
Specific Gravity 1
Solubility Information Solubility in water: miscible.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.