Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Buffer Solution, for Water Hardness, ASTM Formulation, Ricca Chemical

Click to view available options
Quantity:
500 mL
1 L
Specifications
Specifications
Color | Clear colorless to pale-yellow |
pH Range | ∼ 12.5 |
Volume (Metric) | 500 mL |
Packaging | Natural Poly Bottle |
Quantity | 500 mL |
Type | Buffer Solution |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction