Learn More
3M™ Buffered Peptone Water Broth ISO BPW500, Sold by AquaPhoenix Scientific
SCI_ED
Catalog No. S26865 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Quantity:
500 g
Catalog No. Quantity
S26865 500 g
1 options
Catalog No. S26865 Supplier 3M™ Supplier No. BPW500
Add to Cart
Add to Cart
For use in detecting Salmonella and E. coli O157 (including H7)
  • Lowers enrichment times to provide faster time to result with improved growth and recovery of organisms
  • Compliant to ISO 6579 formulation
  • Validated for use with 3M™ molecular detection assay Salmonella and E. coli O157 (including H7)
Specifications
Certifications/Compliance ISO 6579
Detects Salmonella and E. coli O157 (including H7)
Product Type Microbiology Media
For Use With (Equipment) 3M™ molecular detection assay Salmonella and E. coli O157
Quantity 500 g
Tested For Salmonella and E. coli O157
Weight (Metric) 500 g