Learn More
Thermo Scientific™ Buffered Peptone Water (ISO) (Dehydrated)
Catalog No. CM1049T
Change view
Click to view available options
Quantity:
10 kg
25 kg
5 kg
500 g
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CM1049T 5 kg
CM1049B 500 g
CM1049G 10 kg
NC3397093 25 kg
CM1049R Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. CM1049T Supplier Thermo Scientific™ Supplier No. CM1049T
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Pre-enrich food samples prior to the isolation of Salmonella species with this non-selective medium while adhering to ISO 11133:2014
Pre-enrich food samples prior to the isolation of Salmonella species using Thermo Scientific™ Oxoid™ Buffered Peptone Water (ISO) (Dehydrated). Buffered Peptone Water is a non-selective pre-enrichment medium and is tested in accordance with ISO 65791 and ISO 11133:20142. It also provides conditions for resuscitation of cells that have been injured by processes of food preservation.
  • Pre-enrich food samples prior to isolation of Salmonella species
  • Allow high resuscitation of sub-lethally injured cells with medium
    • Low numbers and/or sub-lethally injured Salmonella spp. cells that may be in test food sample
  • Supports high growth and recovery rate, rich in nutrients
  • Prevents bacterial damage due to changes in medium pH
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ISO 6579:2002
Form Powder
Product Type Buffered Peptone Water
For Use With (Application) Allows cells time to repair and multiply before being introduced into a selective culture, Improving chances of recovering Salmonella species from the sample
Quantity 5 kg