Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Buffered Peptone Water (ISO) (Dehydrated)

Catalog No. NC3397093
Change view
Click to view available options
Quantity:
10 kg
25 kg
5 kg
500 g
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NC3397093 25 kg
CM1049B 500 g
CM1049G 10 kg
CM1049T 5 kg
CM1049R Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. NC3397093 Supplier Thermo Scientific™ Supplier No. CM1049K
Add to Cart
Add to Cart
Request a Quote

Pre-enrich food samples prior to the isolation of Salmonella species with this non-selective medium while adhering to ISO 11133:2014

Pre-enrich food samples prior to the isolation of Salmonella species using Thermo Scientific™ Oxoid™ Buffered Peptone Water (ISO) (Dehydrated). Buffered Peptone Water is a non-selective pre-enrichment medium and is tested in accordance with ISO 65791 and ISO 11133:20142. It also provides conditions for resuscitation of cells that have been injured by processes of food preservation.

Low numbers and/or sub-lethally injured Salmonella cells may be present in a test food sample. Pre-enrichment in non-selective Buffered Peptone Water allows the cells time to repair and multiply before being introduced into selective culture, improving the chances of recovering Salmonella species from the sample.

  • Supports high growth and recovery rate: rich in nutrients, the medium produces high resuscitation rates and supports intense growth.
  • Helps recovery of sub-lethally damaged salmonellae: the phosphate buffer system of the medium prevents bacterial damage due to changes in pH of the medium and helps the recovery of sub-lethally damaged salmonellae that may be sensitive to low pH.
  • ISO compliant: tested in accordance with ISO 65791and ISO 11133:20142.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Form Powder
Product Type Pre-enrichment Medium
Quantity 25 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.