Learn More
StatLab™ Buffered Zinc Formalin Prefilled Vial

Description
- Especially recommended for use in immunohistochemistry, as zinc formalin preserves more immunoreactivity than standard NBF
- Vials are prefilled with 10 mL of Buffered Zinc Formalin
Specifications
Specifications
| Product Type | Pre-filled Zinc Formalin Container |
| Closure Included | Yes |
| Graduated | Yes |
| Material | Soda Lime Glass |
| Shape | Cylindrical |
| Sterility | Non-sterile |
| Autoclavable | Not Autoclavable |
| Capacity (Metric) | 20 mL |
| Color | Clear |
| No. per Case | 960 Vials/Cs. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.