Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Buffers for MagJET™ mRNA Enrichment Kit

Purify mRNA from total RNA samples with oligo (dT) poly A+-binding magnetic beads with up to 10-fold enrichment from 5-100 μg RNA input

Specifications

For Use With (Equipment) MagJET mRNA Enrichment Kit
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
FERR2681
View Documents
Thermo Scientific™
R2681
220mL N/A N/A N/A
Description

Description

The MagJET mRNA Enrichment Kit is designed for fast and efficient purification of mRNA from total RNA samples. The oligo (dT) beads bind poly A+ RNA specifically and with high capacity, which ensures maximum extraction of mRNA from a wide range of starting material amount.
The kit utilizes paramagnetic bead technology enabling high yields and robust performance. High binding capacity, uniform particle size, and rapid magnetic response of MagJET oligo (dT) beads makes the technology ideal for high throughput automated nucleic acid purification, as well as for manual nucleic acid purification by low sample throughput users.
The MagJET mRNA Enrichment Kit protocol results in the highest amounts of intact full length RNA transcripts and low rRNA contamination.The resulting high quality purified mRNA is free of proteins, nucleases and other contaminants or inhibitors.

Highlights

  • Up to 10-fold mRNA enrichment
  • Full length high integrity mRNA isolation
  • High yields from broad (5 - 100 μg) RNA input range
  • Compatible with high throughput automation
Applications
Isolated RNA transcripts can be used in:
  • RT-PCR
  • RT-qPCR
Includes
  • Hybridization Buffer for MagJET mRNA Enrichment Kit
  • MagJET Oligo(dT) Beads
  • Wash Buffer for MagJET mRNA Enrichment Kit
  • Water, nuclease-free

Hazardous:No
Quality Control:Thermo Scientific MagJET mRNA Enrichment Kit is qualified by isolating mRNA from 30 μg of total HeLa RNA following the protocol outlined in the manual. The quality of isolated mRNA is determined by measuring rRNA contamination using RT-qPCR. The amount of rRNA is <0.5% from starting material.
Shelf Life:36 Months
Shipping Condition:Ambient
Storage Condition:4 C

Specifications

Specifications

MagJET mRNA Enrichment Kit
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store at 4°C

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.