Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ BugBuster™ HT Protein Extraction Reagent

Catalog No. p-7060190 Shop All MilliporeSigma Products
Click to view available options
Quantity:
100mL
500mL
1L

Eliminates common bioprocessing problems resulting from traditional cell lysis procedures

  • Combines BugBuster Protein Extraction Reagent and Benzonase™ Nuclease in one convenient reagent
  • Soluble proteins are gently extracted from E. coli without exposure to heat or oxidative damage
  • Viscosity is eliminated by nucleic acid digestion in a single step
  • The resulting protein extract can easily be fractionated by conventional purification techniques
  • Ideally suited for application in high throughput protein purifications
TRUSTED_SUSTAINABILITY

Specifications

Quantity 100mL
Content And Storage Store at +15° to +30°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.