Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ BugBuster™ Plus Lysonase™ Kit
Shop All MilliporeSigma Products

Click to view available options
Kit Contents:
BugBuster Protein Extraction Reagent (100mL), Lysonase Bioprocessing Reagent (200μL)
BugBuster Protein Extraction Reagent (500mL), Lysonase Bioprocessing Reagent (1mL)
Description
- The BugBuster and rLysozyme efficiently lyse the bacterial cells while the Benzonase degrades nucleic acids to facilitate downstream processing
- Use 5mL BugBuster and 10μL Lysonase per 1g cell paste
- The kits provide sufficient reagents for protein extraction from 20g and 100g cell paste
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Quantity | 1 kit |
| Kit Contents | BugBuster Protein Extraction Reagent (500mL), Lysonase Bioprocessing Reagent (1mL) |
| Content And Storage | Multiple storage requirements |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction