Learn More
Poltex Bulk Bin, 8 in. Wide Comp
Description
Simple Bulk Bin with open top for filling and front for dispensing.
Durable and cleanable PETG construction.
Choose from:
- Wall/countertop mount with rubber bumpons on base
- VHB tape mount (very high bond tape included)
- Magnet mount (powerful magnets included)
Specifications
Specifications
| Type | Dispensing Bin |
| Material | PETG |
| Color | Clear |
| Shape | Square |
| Mounting Type | Tape Mount |
| Autoclavable | Not Autoclavable |