Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poltex Bulk Conical Tube Bin
SDP

Catalog No. p-7163850
Click to view available options
Mounting:
Keyhole slots for wall mount
Powerful Magnet Mount
Powerful VHB Tape Mount

Bulk tube bin to help organize hood and/or bench

Specifications

Includes Bin
Autoclavable Not Autoclavable
Color Clear
Holds Bulk Conical Tubes, Test Tubes, Microcentrifuge Tubes
Material PETG
Positions 1
Sterility Non-sterile
For Use With (Equipment) Bulk conical tubes, test tubes, microcentrifuge tubes
Mounting Powerful Magnet Mount
Quantity 1
Length (Metric) 205 mm
Width (Metric) 185 mm
Height (Metric) 221 mm
Weight (Metric) 250 g
Show More Show Less

Consumables not included.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.