Learn More
Bullard™ 3000R Series Safety Cap
SDP
Classic cap-style hard hat
$32.95 - $39.00
Specifications
Suspension Type 6-Point Ratchet
View More Specs
Products 3
Catalog Number Mfr. No. Color Price Quantity & Availability  
Catalog Number Mfr. No. Color Price Quantity & Availability  
19-052684
SDP
View Documents
Bullard™
30RDR
Red
Each for $32.95
Case of 20 EA for $627.00
Only null left
Add to Cart
19-086337
SDP
View Documents
Bullard™
30TNR
Tan
Each for $39.00
Only null left
Add to Cart
19-071234
SDP
View Documents
Bullard™
30WHRU
White N/A N/A N/A
Description
Description
  • Six-point adjustable Flex-Gear™ suspension with seamless 1 in. nylon crown straps
  • Vertical height adjustment provides custom fit
  • Replaceable absorbent cotton brow pad, a nape cushion, and a large, easily gripable sizing knob
  • Accessory slots accept most attachments
Compliance
Meets ANSI Z89.1-2003, Type 1, Class E and G
Specifications
Specifications
6-Point Ratchet
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.