Learn More
Bullard™ 41 Series Breathing AirLine Filter Replacement Parts
SDP
Catalog No. 19824401 Shop All Bullard Company Products
Change view
Click to view available options
Product Type:
Filter Cartridge
Manifold Assembly
Model:
41AF
41P2M
41P6UPK
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Model
19-824-401 Manifold Assembly 41P2M
19-021409 Filter Cartridge 41AF
19-824-403 Manifold Assembly 41P6UPK
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
The 41 Series Airline filters do not remove carbon monoxide and other toxic gases. Filter cartridge must be changed periodically for maximum efficiency. Frequency of cartridge changes depends on operating conditions. Cartridge should be changed immediately if respirator wearer feels, smells or tastes contaminants inside the respirator.
Catalog No. 19-824-401 Supplier Bullard™ Supplier No. 41P2M
This item is not returnable. View return policy
The 41 Series Airline filters do not remove carbon monoxide and other toxic gases. Filter cartridge must be changed periodically for maximum efficiency. Frequency of cartridge changes depends on operating conditions. Cartridge should be changed immediately if respirator wearer feels, smells or tastes contaminants inside the respirator.
Replacement parts for use with 41 Series breathing airline filters.
  • Replacement parts for 41 Series airlines
  • Filter cartridge
  • 6-outlet manifold upgrade for 41P6 includes:
    • Manifold
    • Regulator assembly
    • Relief valve
    • Gauge
  • 2-outlet manifold assembly for 41P2 and 41P2E includes:
    • Manifold with regulator
    • Relief valve
    • Gauge
Specifications
For Use With (Equipment) 41 Series Airline Filter
Includes Manifold, regulator, relief valve, and gauge
Model 41P2M
Product Type Manifold Assembly