Learn More
Bullard™ 41 Series Breathing AirLine Filter Replacement Parts
Replacement parts for use with 41 Series breathing airline filters.
Supplier: Bullard™ 41P6UPK
Alert:
The 41 Series Airline filters do not remove carbon monoxide and other toxic gases. Filter cartridge must be changed periodically for maximum efficiency. Frequency of cartridge changes depends on operating conditions. Cartridge should be changed immediately if respirator wearer feels, smells or tastes contaminants inside the respirator.
Description
- Replacement parts for 41 Series airlines
- Filter cartridge
- 6-outlet manifold upgrade for 41P6 includes:
- Manifold
- Regulator assembly
- Relief valve
- Gauge
- 2-outlet manifold assembly for 41P2 and 41P2E includes:
- Manifold with regulator
- Relief valve
- Gauge
Specifications
41 Series Airline Filter | |
Manifold, regulator, relief valve, and gauge | |
41P6UPK | |
Manifold Assembly |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.