Learn More
Bullard™ Inlet and Outlet Filters for Electric Free-Air™ Pumps
SDP
Catalog No. p-58563 Shop All Bullard Company Products
Change view
Click to view available options
Product Type:
Inlet filter
Outlet filter
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
19-021308 Outlet filter
19-021342 Inlet filter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-021308 Supplier Bullard™ Supplier No. S17101
Add to Cart
Add to Cart
For use with Free-Air pumps
Coupler Splitter/Adapter: Converts coupler to double 1/2 in. Industrial Interchange female couplers (one flow through, one shut-off)
Inlet Filter: Change every 500 hours or as required
Outlet Filter: Carbofine; change every 200 hours or as required
Specifications
Model S17101
Product Type Outlet filter