Learn More
Bullard™ Blowers for EVA™ Powered-Air Purifying Respirators
Shop All Bullard Company Products
Click to view available options
Includes:
Blower, battery, airflow indicator, single charger
Blower, battery; No charger or filter
Specifications
Specifications
Battery Type | Lithium |
Certifications/Compliance | NIOSH |
Connection Type | Belt |
Product Type | Blower Assembly |
For Use With (Equipment) | Eva™ powered air purifying respirator |
Includes | Blower, battery, airflow indicator, single charger |
Safety Services
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.