Learn More
Bullard™ Flow Control Assemblies for CC20 Series Airline Respirator
SDP
Catalog No. 19824384 Shop All Bullard Company Products
Click to view available options
Material:
Brass
Stainless Steel
Constant flow fittings for CC20 Series airline respirators.
  • Airline respirator accessory
  • Constant flow fitting
  • For use with CC20
  • 0.25 in. industrial interchange
Specifications
Material Brass
Product Type Constant Flow Control Valve
For Use With (Equipment) CC20 Series Compressed Air Respirator
Includes Industrial interchange (Hansen-compatible) quick-disconnect nipple, nylon belt, threaded hose connector
Model F30B
Diameter (English) Interior 0.25 in.
Diameter (Metric) Interior 0.63 cm
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.