Learn More
Bullard™ Free-Air EDP30 Electric Driven Pumps
SDP
Catalog No. 17604207 Shop All Bullard Company Products
Catalog No. 17-604-207 Supplier Bullard™ Supplier No. EDP30B460
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Supports climate control devices
Shipped complete with inlet and outlet filter cartridges, pressure gauges, pressure relief valves, and one air supply hose quick-disconnect pod with six 0.5 in. Industrial Interchange (Hansen compatible) quick-disconnect couplers.
  • Supplies air for four hood-style, six half or full face mask airline respirators, or two hood-style respirators equipped with Frigitron climate control devices
  • Respirators must be approved to operate on 30 psig or less
  • Easy operation and low maintenance
  • Completely mobile and portable
  • No expensive installation costs
Specifications
Applications Support Climate Control Devices
Includes Inlet and outlet filter cartridge, pressure gauges, pressure relief valves and one air supply hose quick-disconnect pod with six 0.5 in. industrial interchange quick-disconnect couplers, reversing starter
Product Type Electric Driven Air Pump