Learn More
Bullard™ Free-Air EDP50 Electric Driven Pumps
SDP
Catalog No. 17604213 Shop All Bullard Company Products
Catalog No. 17-604-213 Supplier Bullard™ Supplier No. EDP50B230
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Supports up to ten respirators
Operates without temperature alarms, CO monitors or airline filters.
  • Supplies air for seven hood-style or ten half or full face mask airline respirators
  • Features point of attachment, quick disconnect pods that can be extended up to 300 ft. from pump
  • Air flow of 50 cfm at 5 psig
  • Easy operation and low maintenance
  • Completely mobile and portable
  • No expensive installation costs
Specifications
Includes Inlet and outlet filter cartridge, pressure gauges, pressure relief valves and one air supply hose quick-disconnect pod with six 0.5 in. industrial interchange quick-disconnect couplers
Product Type Electric Driven Air Pump