Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bullard™ Standard Series S51 Caps
Shop All Bullard Company Products

Click to view available options
Color:
Black
Blue
Charcoal Brown
Forest Green
Gray
Hi-vis Green
Hi-vis Orange
Hi-vis Yellow
Kelly Green
Navy Blue
Orange
Pacific Blue
Red
Tan
White
Yellow
Suspension Type:
4-Point Pin-Lock
4-Point Ratchet
Flex-Gear Ratchet
Standard
Description
- Constructed of polyethylene
- Soft poly browpad absorbs moisture
- Rain trough provides extra protection from snow, rain, dust and dirt
- Accessory slots accommodate a variety of optional attachments
- Available with standard four-point suspension or four-point Flex-Gear™ ratchet suspension featuring two vertical adjustments
- Wide 0.75 in. (1.9cm) crown straps and low nape strap keep caps securely in place
Order Info
Note: There is a minimum order quantity of 20.
Compliance
ANSI/ISEA Z89.1-2009, Type I, Class E & G and CSA Z94.1-2005, Type 1, Class E & G requirements.
Specifications
Specifications
| Certifications/Compliance | Meets or exceeds ANSI Z891-1997; Type 1; Class E G and C |
| Color | Hi-vis Green |
| Suspension Type | 4-Point Ratchet |
Safety Services
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction
