Learn More
Bullard™ Standard Series S62 Vented Hardhats
SDP
Catalog No. 19160465 Shop All Bullard Company Products
Change view
Click to view available options
Color:
Black
Blue
Charcoal Brown
Dove Gray
Forest Green
Hi-vis Orange
Hi-vis Yellow
Kelly Green
Kentucky Blue
Orange
Pacific Blue
Red
Tan
White
Yellow
Suspension Type:
4-Point Flex-Gear Ratchet
4-Point Pin-Lock
Flex-Gear Ratchet
23 product options available for selection
Product selection table with 23 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Suspension Type
19-160-465 Hi-vis Yellow 4-Point Pin-Lock
19-160-447 Kentucky Blue 4-Point Flex-Gear Ratchet
19-160-448 Yellow 4-Point Flex-Gear Ratchet
19-160-449 Charcoal Brown Flex-Gear Ratchet
19-160-452 Kelly Green Flex-Gear Ratchet
19-160-450 Orange 4-Point Flex-Gear Ratchet
19-160-455 Tan 4-Point Flex-Gear Ratchet
19-160-453 Dove Gray Flex-Gear Ratchet
19-160-456 Black Flex-Gear Ratchet
19-160-454 Pacific Blue Flex-Gear Ratchet
19-160-443 White 4-Point Flex-Gear Ratchet
19-160-445 Orange Flex-Gear Ratchet
19-160-458 Red 4-Point Pin-Lock
19-160-459 Orange 4-Point Pin-Lock
19-160-461 Blue 4-Point Pin-Lock
19-160-460 Forest Green 4-Point Pin-Lock
19-160-464 Hi-vis Orange 4-Point Pin-Lock
19-160-463 Charcoal Brown 4-Point Pin-Lock
19-160-468 Pacific Blue 4-Point Pin-Lock
19-160-466 Kelly Green 4-Point Pin-Lock
19-160-467 Dove Gray 4-Point Pin-Lock
19-160-469 Tan 4-Point Pin-Lock
19-160-470 Black 4-Point Pin-Lock
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 23 options available.
23 options
Catalog No. 19-160-465 Supplier Bullard™ Supplier No. 62HYP
Low-profile shell design with four-point suspension system
Setting the Standard. Sleek, low-profile design with a patented Flex-Gear ratchet suspension option.
Features
  • Absorbent, replaceable cotton brow pad is pillowed for maximum comfort
  • Ideal for construction and everyday work environments
  • One of the lightest helmets of its class
  • Vented, providing increased ventilation and comfort
  • Accessory slots allow attachment of accessories such as earmuffs and face protection
  • Vertical height adjustment capability (2 front, 2 rear)
Pinlock or Ratchet Suspension
  • Choose 4-point pinlock suspension or 4-point Flex-Gear ratchet suspension
  • Pinlock suspension provides incremental sizing of 1/16 hat size (5mm), for a secure fit
  • Ratchet suspension can be adjusted between hat sizes 6 1/2-8
Order Info
Snap-on replacement brow pads are available (see below).
Compliance
ANSI Z89.1-2003, Type I, Class E and G (S61 Model); Class C (S62 Vented).
Specifications
Certifications/Compliance ANSI Z89.1, Type I, Class C
Color Hi-vis Yellow
Suspension Type 4-Point Pin-Lock
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.