Learn More
Bullard™ Standard Series S62 Vented Hardhats
SDP
Catalog No. 19160443 Shop All Bullard Company Products
Click to view available options
Color:
Black
Blue
Charcoal Brown
Dove Gray
Forest Green
Hi-vis Orange
Hi-vis Yellow
Kelly Green
Kentucky Blue
Orange
Pacific Blue
Red
Tan
White
Yellow
Suspension Type:
4-Point Flex-Gear Ratchet
4-Point Pin-Lock
Flex-Gear Ratchet
This item is not returnable. View return policy
This item is not returnable. View return policy
Low-profile shell design with four-point suspension system
Setting the Standard. Sleek, low-profile design with a patented Flex-Gear ratchet suspension option.
Features
  • Absorbent, replaceable cotton brow pad is pillowed for maximum comfort
  • Ideal for construction and everyday work environments
  • One of the lightest helmets of its class
  • Vented, providing increased ventilation and comfort
  • Accessory slots allow attachment of accessories such as earmuffs and face protection
  • Vertical height adjustment capability (2 front, 2 rear)
Pinlock or Ratchet Suspension
  • Choose 4-point pinlock suspension or 4-point Flex-Gear ratchet suspension
  • Pinlock suspension provides incremental sizing of 1/16 hat size (5mm), for a secure fit
  • Ratchet suspension can be adjusted between hat sizes 6 1/2-8
Order Info
Snap-on replacement brow pads are available (see below).
Compliance
ANSI Z89.1-2003, Type I, Class E and G (S61 Model); Class C (S62 Vented).
Specifications
Certifications/Compliance ANSI Z89.1, Type I, Class C
Color White
Size Standard
Suspension Type 4-Point Flex-Gear Ratchet
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.