Description
Setting the Standard. Sleek, low-profile design with a patented Flex-Gear ratchet suspension option.
Features
- Absorbent, replaceable cotton brow pad is pillowed for maximum comfort
- Ideal for construction and everyday work environments
- One of the lightest helmets of its class
- Vented, providing increased ventilation and comfort
- Accessory slots allow attachment of accessories such as earmuffs and face protection
- Vertical height adjustment capability (2 front, 2 rear)
- Choose 4-point pinlock suspension or 4-point Flex-Gear ratchet suspension
- Pinlock suspension provides incremental sizing of 1/16 hat size (5mm), for a secure fit
- Ratchet suspension can be adjusted between hat sizes 6 1/2-8
Order Info
Snap-on replacement brow pads are available (see below).
Compliance
ANSI Z89.1-2003, Type I, Class E and G (S61 Model); Class C (S62 Vented).
Specifications
Specifications
| Certifications/Compliance | ANSI Z89.1, Type I, Class C |
| Color | Orange |
| Suspension Type | Flex-Gear Ratchet |
Safety Services
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.