Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bulwark™ FR Men's Midweight Excel FR™ Comfortouch™ Flame Resistant Deluxe Insulated Bib Overall

Catalog No. 19080323 Shop All Red Kap Industries Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Style:
Long
Regular
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Style
19-080-323 2X-Large Regular
19-080-319 Small Regular
19-080-322 X-Large Regular
19-080-320 Medium Regular
19-080-321 Large Regular
19-080-324 3X-Large Regular
19-080-325 Large Long
19-080-327 2X-Large Long
19-080-326 X-Large Long
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Catalog No. 19-080-323 Supplier Bulwark™ FR Supplier No. BLC8NV RG XXL
Add to Cart
Add to Cart
This item is not returnable. View return policy

For cold jobs, this Midweight Insulated Bib Overall offers unbeatable warmth with CAT 3 AR/FR protection and deluxe features like rugged, double-reinforced knees. Large patch pockets and bonus utility pockets ensure there's a place for all of your tools.

  • Certifications Level 1: NFPA 2112, NFPA 70E
  • Certifications Level 2: Category 3 Protection
  • Hazard:
  • Primary Closure: Concealed Zipper
  • Wash Care: Industrial Laundry - Heavy Soil, Industrial Laundry - Light Soil, Home Wash
  • Fabric: Shell: Flame resistant, 7 oz. (235 g/m2) Twill, Water-repellent, Insulation Package: 14 oz. (474 g/m2)
  • Blend: Shell: 88% Cotton/12% Nylon, Insulation Package: 50% Modacrylic/50% Cotton
  • Closure: Elasticized shoulder straps with non-conductive adjustable hardwear
  • Pocket: Five large patch pockets and hip pocket with closure
  • Features:
    • Reinforced double knees
    • Elasticized shoulder straps with non-conductive adjustable hardwear
  • Product Collection: Excel FR Comfortouch
  • Protection: ATPV Arc Rating: 36
  • Country of Origin: Imported, Made in USA Fabric
  • Fit Details: Designed to be worn over other clothing. Fit over work boots.
  • Gender: Male
  • Materials: Cotton/Nylon
  • Brand: Bulwark™

Specifications

Closure Type Hip Zipper with Concealed Snap
Color Navy
Material Cotton, Nylon
Size 2X-Large
Construction Cotton, Nylon, Modacrylic lining
For Use With (Application) Excellent barrier to dry particles, microorganisms and non-hazardous liquids
Length (English) Inseam 31.5 in.
Length (Metric) Inseam 80 cm
No. of Pockets 5
Size (English) Chest 48.5 to 50.5 in.
Size (English) Waist 48.5 to 50.5 in.
Size (Metric) Chest 123.1 to 128.2 cm
Size (Metric) Waist 123.1 to 128.2 cm
Style Regular
Waist Style Elastic
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.