Learn More
Bulwark™ FR Men's Midweight Excel FR™ Comfortouch™ Flame Resistant Deluxe Insulated Bib Overall
Shop All Red Kap Industries ProductsDescription
- Certifications Level 1: NFPA 2112, NFPA 70E
- Certifications Level 2: Category 3 Protection
- Hazard:
- Primary Closure: Concealed Zipper
- Wash Care: Industrial Laundry - Heavy Soil, Industrial Laundry - Light Soil, Home Wash
- Fabric: Shell: Flame resistant, 7 oz. (235 g/m2) Twill, Water-repellent, Insulation Package: 14 oz. (474 g/m2)
- Blend: Shell: 88% Cotton/12% Nylon, Insulation Package: 50% Modacrylic/50% Cotton
- Closure: Elasticized shoulder straps with non-conductive adjustable hardwear
- Pocket: Five large patch pockets and hip pocket with closure
- Features:
- Reinforced double knees
- Elasticized shoulder straps with non-conductive adjustable hardwear
- Product Collection: Excel FR Comfortouch
- Protection: ATPV Arc Rating: 36
- Country of Origin: Imported, Made in USA Fabric
- Fit Details: Designed to be worn over other clothing. Fit over work boots.
- Gender: Male
- Materials: Cotton/Nylon
- Brand: Bulwark™
Specifications
Specifications
| Closure Type | Hip Zipper with Concealed Snap |
| Color | Navy |
| Material | Cotton, Nylon |
| Size | Large |
| Construction | Cotton, Nylon, Modacrylic lining |
| For Use With (Application) | Excellent barrier to dry particles, microorganisms and non-hazardous liquids |
| Length (English) Inseam | 33.5 in. |
| Length (Metric) Inseam | 85 cm |
| No. of Pockets | 5 |
| Size (English) Chest | 40.5 to 42.5 in. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.