Learn More
Bulwark™ FR Men's Heavyweight Excel FR™ Comfortouch™ Flame Resistant Deluxe Insulated Bib Overall
Shop All Red Kap Industries ProductsDescription
- Primary Closure: Zipper
- Wash Care: Industrial Laundry - Heavy Soil, Industrial Laundry - Light Soil, Home Wash
- Fabric: Shell: Flame resistant, 11 oz. (390 g/m2) Duck, Water repellent
- Blend: Shell: 88% Cotton/12% Nylon, Lining: 100% Cotton, Insulation: 100% Modacrylic
- Closure: Elasticized shoulder straps with non-conductive adjustable hardware
- Pocket: One large front bib pocket with pencil stall and two set-in front swing pockets, One watch pocket and two large hip pockets, Two-piece rule pocket and one tool pocket
- Features
- Reinforced double knees
- One large front bib pocket with pencil stall
- Two set-in front swing pockets
- Elasticized shoulder straps with non-conductive adjustable hardware
- Product Collection: Excel FR Comfortouch, Canvas
- Protection: ATPV Arc Rating: 44
- Country of Origin: Imported, Made in USA Fabric
- Fit Details: Designed to be worn over other clothing. Fit over work boots.
- Gender: Male
- Materials: Cotton/Nylon
- Brand: Bulwark™
Specifications
Specifications
| Closure Type | Hip Zipper with Concealed Snap |
| Color | Brown |
| Material | Cotton, Nylon |
| Size | Small |
| Construction | Cotton, Nylon, Modacrylic lining |
| For Use With (Application) | Excellent barrier to dry particles, microorganisms and non-hazardous liquids |
| Length (English) Inseam | 31.5 in. |
| Length (Metric) Inseam | 80 cm |
| No. of Pockets | 8 |
| Size (English) Chest | 32.5 to 34.5 in. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.