Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bulwark™ FR Men's Heavyweight Excel FR™ Comfortouch™ Flame Resistant Deluxe Insulated Bib Overall

Catalog No. 19080311 Shop All Red Kap Industries Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Style:
Long
Regular
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Style
19-080-311 Medium Regular
19-080-313 X-Large Regular
19-080-310 Small Regular
19-080-312 Large Regular
19-080-314 2X-Large Regular
19-080-315 3X-Large Regular
19-080-316 Large Long
19-080-317 X-Large Long
19-080-318 2X-Large Long
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Catalog No. 19-080-311 Supplier Bulwark™ FR Supplier No. BLN4BD RG M
Add to Cart
Add to Cart
This item is not returnable. View return policy

If you're looking for the ultimate in rugged durability, warmth, and FR protection, then this Heavyweight Brown Duck Bib Overall is made for you. With CAT 4 protection, insulation throughout and side vent openings, these overalls with keep you warm and comfortable while they protect you.

  • Primary Closure: Zipper
  • Wash Care: Industrial Laundry - Heavy Soil, Industrial Laundry - Light Soil, Home Wash
  • Fabric: Shell: Flame resistant, 11 oz. (390 g/m2) Duck, Water repellent
  • Blend: Shell: 88% Cotton/12% Nylon, Lining: 100% Cotton, Insulation: 100% Modacrylic
  • Closure: Elasticized shoulder straps with non-conductive adjustable hardware
  • Pocket: One large front bib pocket with pencil stall and two set-in front swing pockets, One watch pocket and two large hip pockets, Two-piece rule pocket and one tool pocket
  • Features
    • Reinforced double knees
    • One large front bib pocket with pencil stall
    • Two set-in front swing pockets
    • Elasticized shoulder straps with non-conductive adjustable hardware
  • Product Collection: Excel FR Comfortouch, Canvas
  • Protection: ATPV Arc Rating: 44
  • Country of Origin: Imported, Made in USA Fabric
  • Fit Details: Designed to be worn over other clothing. Fit over work boots.
  • Gender: Male
  • Materials: Cotton/Nylon
  • Brand: Bulwark™

Specifications

Closure Type Hip Zipper with Concealed Snap
Color Brown
Material Cotton, Nylon
Size Medium
Construction Cotton, Nylon, Modacrylic lining
For Use With (Application) Excellent barrier to dry particles, microorganisms and non-hazardous liquids
Length (English) Inseam 31.5 in.
Length (Metric) Inseam 80 cm
No. of Pockets 8
Size (English) Chest 36.5 to 38.5 in.
Size (English) Waist 36.5 to 38.5 in.
Size (Metric) Chest 92.7 to 97.7 cm
Size (Metric) Waist 92.7 to 97.7 cm
Style Regular
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.