Learn More
Bulwark™ FR Men's Midweight Excel FR™ Comfortouch™ Work Shirt, Navy
Shop All Red Kap Industries ProductsDescription
- Certifications Level 1: ASTM F1506, ANSI 107, NFPA 70E
- Certifications Level 2: Category 2 Protection
- Hazard: 2
- Primary Closure: Button
- Wash Care: Industrial Laundry - Heavy Soil, Industrial Laundry - Light Soil, Home Wash
- Fabric: Flame resistant, 7 oz. (235 g/m2)
- Blend: 88% Cotton/12% Nylon
- Closure: Button-down front with one-piece lined cuffs with button closures
- Collar: Lay-flat collar
- Pocket: Two chest pockets with button flap closures and sewn-in pencil stall
- Features: Hemmed front with button closures
- Product Collection: Excel FR Comfortouch
- Protection: ATPV Arc Rating: 8.6
- Country of Origin: Imported, Made in USA Fabric
- Fit Details: Regular Fit
- Gender: Male
- Materials: Cotton/Nylon
- Brand: Bulwark™
Specifications
Specifications
| Size | 3X-Large |
| Gender | Men's |
| Material | Cotton, Nylon |
| Color | Navy |
| Closure Type | Button |
| Hazard Risk Category | 2 |
| Arc Flash Rating | ATPV 8.6 Calories/cm2 |
| Certifications/Compliance | ASTM F1506, ANSI 107, NFPA 70E, Category 2 Protection |
| Weight (English) | 7 oz. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.