Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bulwark™ FR Women's Midweight Excel FR™ Comfortouch™ Flame Resistant Work Pant, Khaki

Catalog No. 17150590 Shop All Red Kap Industries Products
Change view
Click to view available options
Size (English) Inseam:
28 in.
30 in.
32 in.
34 in.
Size:
4
6
8
10
12
14
16
18
20
22
24
44 product options available for selection
Product selection table with 44 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size (English) Inseam Size
17-150-590 28 in. 16
17-150-566 28 in. 4
17-150-567 30 in. 4
17-150-568 32 in. 4
17-150-569 34 in. 4
17-150-570 28 in. 6
17-150-571 30 in. 6
17-150-572 32 in. 6
17-150-573 34 in. 6
17-150-574 28 in. 8
17-150-575 30 in. 8
17-150-576 32 in. 8
17-150-578 28 in. 10
17-150-582 28 in. 12
17-150-577 34 in. 8
17-150-579 30 in. 10
17-150-580 32 in. 10
17-150-581 34 in. 10
17-150-583 30 in. 12
17-150-584 32 in. 12
17-150-585 34 in. 12
17-150-591 30 in. 16
17-150-588 32 in. 14
17-150-589 34 in. 14
17-150-586 28 in. 14
17-150-587 30 in. 14
17-150-592 32 in. 16
17-150-593 34 in. 16
17-150-594 28 in. 18
17-150-595 30 in. 18
17-150-600 32 in. 20
17-150-597 34 in. 18
17-150-596 32 in. 18
17-150-599 30 in. 20
17-150-601 34 in. 20
17-150-598 28 in. 20
17-150-602 28 in. 22
17-150-608 32 in. 24
17-150-603 30 in. 22
17-150-605 34 in. 22
17-150-609 34 in. 24
17-150-604 32 in. 22
17-150-606 28 in. 24
17-150-607 30 in. 24
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 44 options available.
44 options
This item is not returnable. View return policy
Catalog No. 17-150-590 Supplier Bulwark™ FR Supplier No. PLW3KHB 16 28

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

This NFPA™ 2112 compliant Midweight CAT 2 AR/FR Work Pant offers a classic look that will keep you comfortable and FR protection to keep you safe. Designed especially for women, with the right fit and sizing.

  • Wash Care: Industrial Laundry - Heavy Soil, Industrial Laundry - Light Soil, Home Wash
  • Fabric: Westex Ultrasoft™ Flame resistant, 9 oz. (305 g/m2) Twill
  • Blend: 88% Cotton/12% Nylon
  • Closure: Interlined waistband with button closure
  • Pocket: Two slack-style front pockets, two set-in hip pockets and left hip with button closure
  • Features:
    • Interlined waistband with button closure
    • Two slack style front pockets
    • Two set-in hip pockets
    • Left hip pocket with button closure
  • Product Collection: Excel FR Comfortouch
  • Protection: ATPV Arc Rating: 12
  • Country of Origin: Imported, Made in USA Fabric
  • Fit Details: Sits at the waist. Regular in the seat and thigh. Slightly tapered leg.
  • Gender: Female
  • Materials: Ultra Soft, Cotton/Nylon
  • Brand: Bulwark™

Specifications

Size 16
Gender Women's
Product Type Flame-resistant Work Pants
Material Cotton, Nylon
Color Navy
No. of Pockets 4
Closure Type Button
Hazard Risk Category HRC2
Arc Flash Rating ATPV 12.2 Calories/cm2
Certifications/Compliance Meets NFPA 2112
Size (English) Inseam 28 in.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.