Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bulwark™ FR Men's Midweight Flame Resistant Pocketless Concealed-Gripper Work Shirt

Catalog No. 18120327 Shop All Red Kap Industries Products
Change view
Click to view available options
Size:
2X-Large
2X-Large (Long)
3X-Large
Large
Large (Long)
Medium
Medium (Long)
Small
X-Large
X-Large (Long)
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18-120-327 Medium
18-120-326 Small
18-120-328 Medium (Long)
18-120-329 Large
18-120-330 Large (Long)
18-120-331 X-Large
18-120-332 X-Large (Long)
18-120-335 3X-Large
18-120-334 2X-Large (Long)
18-120-333 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
This item is not returnable. View return policy
Catalog No. 18-120-327 Supplier Bulwark™ FR Supplier No. SLS2LB RG M
Add to Cart
Add to Cart
This item is not returnable. View return policy

The Men's Midweight Flame Resistant Pocketless Concealed-Gripper Work Shirt is streamlined, pocketless, and FR. This is your go-to work shirt, with CAT 2 AR/FR protection and NFPA™ 2112 compliance.

  • Certifications Level 1: ASTM F1506, NFPA 2112, NFPA 70E
  • Certifications Level 2: Category 2 Protection
  • Hazard:
  • Primary Closure: Snap/Gripper
  • Wash Care: Industrial Laundry - Heavy Soil, Industrial Laundry - Light Soil, Home Wash
  • Fabric: Flame resistant, 7 oz. (235 g/m2)
  • Blend: 88% Cotton/12% Nylon
  • Closure: Concealed gripper-front closure
  • Collar: Lay-flat collar
  • Features: One-piece lined cuffs with closures, Sleeve vents
  • Product Collection: Excel FR Comfortouch
  • Packaging: Hang tag
  • Protection: ATPV Arc Rating: 8.6
  • Country of Origin: Imported, Made in USA Fabric
  • Fit Details: Regular Fit
  • Gender: Male
  • Materials: Cotton/Nylon
  • Brand: Bulwark™

Specifications

Size Medium
Gender Men's
Material Cotton, Nylon
Color Light Blue
Neck Style Collar
Closure Type Hemmed front w/concealed gripper closures
Resistance Flame
Certifications/Compliance ASTM F1506, NFPA 2112, NFPA 70E, Category 2 Protection
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.