Learn More
Bulwark™ FR Men's Midweight Flame Resistant Pocketless Concealed-Gripper Work Shirt
Shop All Red Kap Industries ProductsDescription
- Certifications Level 1: ASTM F1506, NFPA 2112, NFPA 70E
- Certifications Level 2: Category 2 Protection
- Hazard:
- Primary Closure: Snap/Gripper
- Wash Care: Industrial Laundry - Heavy Soil, Industrial Laundry - Light Soil, Home Wash
- Fabric: Flame resistant, 7 oz. (235 g/m2)
- Blend: 88% Cotton/12% Nylon
- Closure: Concealed gripper-front closure
- Collar: Lay-flat collar
- Features: One-piece lined cuffs with closures, Sleeve vents
- Product Collection: Excel FR Comfortouch
- Packaging: Hang tag
- Protection: ATPV Arc Rating: 8.6
- Country of Origin: Imported, Made in USA Fabric
- Fit Details: Regular Fit
- Gender: Male
- Materials: Cotton/Nylon
- Brand: Bulwark™
Specifications
Specifications
| Size | Medium (Long) |
| Gender | Men's |
| Material | Cotton, Nylon |
| Color | Light Blue |
| Neck Style | Collar |
| Closure Type | Hemmed front w/concealed gripper closures |
| Resistance | Flame |
| Certifications/Compliance | ASTM F1506, NFPA 2112, NFPA 70E, Category 2 Protection |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.