Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
- Home
- Products
- Personal Protective Equipment
- Safety Clothing
- Pants, Trousers, and Leggings
- Heat and Fire Resistant Pants, Trousers, and Leggings
- Bulwark™ FR Women's Lightweight Flame Resistant Cargo Pant
Bulwark™ FR Women's Lightweight Flame Resistant Cargo Pant
Catalog No. 17150488 Shop All Red Kap Industries Products
Change view
Click to view available options
Size:
4
6
8
10
12
14
16
18
20
22
24
Size (English) Inseam:
28 in.
30 in.
32 in.
34 in.
44 product options available for selection
| Catalog No. | Size | Size (English) Inseam |
|---|---|---|
| 17-150-488 | 14 | 34 in. |
| 17-200-803 | 22 | 28 in. |
| 17-200-802 | 20 | 34 in. |
| 17-200-804 | 22 | 30 in. |
| 17-200-808 | 24 | 30 in. |
| 17-200-806 | 22 | 34 in. |
| 17-200-807 | 24 | 28 in. |
| 17-200-805 | 22 | 32 in. |
| 17-200-809 | 24 | 32 in. |
| 17-200-810 | 24 | 34 in. |
| 17-150-465 | 4 | 28 in. |
| 17-150-466 | 4 | 30 in. |
| 17-150-470 | 6 | 30 in. |
| 17-150-469 | 6 | 28 in. |
| 17-150-467 | 4 | 32 in. |
| 17-150-474 | 8 | 30 in. |
| 17-150-472 | 6 | 34 in. |
| 17-150-471 | 6 | 32 in. |
| 17-150-473 | 8 | 28 in. |
| 17-150-468 | 4 | 34 in. |
| 17-150-475 | 8 | 32 in. |
| 17-150-476 | 8 | 34 in. |
| 17-150-478 | 10 | 30 in. |
| 17-150-481 | 12 | 28 in. |
| 17-150-477 | 10 | 28 in. |
| 17-150-480 | 10 | 34 in. |
| 17-150-482 | 12 | 30 in. |
| 17-150-479 | 10 | 32 in. |
| 17-150-483 | 12 | 32 in. |
| 17-150-487 | 14 | 32 in. |
| 17-150-485 | 14 | 28 in. |
| 17-150-486 | 14 | 30 in. |
| 17-150-484 | 12 | 34 in. |
| 17-150-489 | 16 | 28 in. |
| 17-150-490 | 16 | 30 in. |
| 17-150-491 | 16 | 32 in. |
| 17-150-493 | 18 | 28 in. |
| 17-150-492 | 16 | 34 in. |
| 17-150-494 | 18 | 30 in. |
| 17-150-497 | 20 | 28 in. |
| 17-150-496 | 18 | 34 in. |
| 17-150-495 | 18 | 32 in. |
| 17-150-498 | 20 | 30 in. |
| 17-150-499 | 20 | 32 in. |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 44 options available.
This item is not returnable.
View return policy
This item is not returnable.
View return policy
Description
- Primary Closure: Hook and Eye
- Wash Care: Industrial Laundry - Light Soil, Home Wash
- Fabric: Flame resistant, 7 oz. (235 g/m2)
- Blend: 47% Modacrylic/37% Lyocell/16% Aramid
- Closure: Waistband with hook-and-bar closure
- Pocket: Two front slack-style pockets and two cargo utility pockets, Left cargo pocket has two snap closures and right utility pocket is open
- Features:
- Waistband with hook-and-bar closure
- Two utility pockets
- Left cargo pocket has two snap closures
- Right utility pocket is open
- Product Collection: Cooltouch 2
- Protection: ATPV Arc Rating: 11
- Country of Origin: Imported, Made in USA Fabric
- Fit Details: Sits at the waist. Regular in the seat and thigh. Slightly tapered leg.
- Gender: Female
- Materials: Modacrylic, Lyocell, Aramid
- Brand: Bulwark™
Specifications
Specifications
| Size | 14 |
| Product Type | Flame-resistant Cargo Pants |
| Gender | Women's |
| Material | Modacrylic, Lyocell, Polyamide-Imide, Aramid |
| Color | Navy |
| No. of Pockets | 5 |
| Closure Type | Button |
| Resistant To | Flame |
| Hazard Risk Category | HRC2 |
| Arc Flash Rating | ATPV 10.1 Calories/cm2 |
| Certifications/Compliance | Meets NFPA 2112 |
| Size (English) Inseam | 34 in. |
| Style | Unhemmed |
| Show More Show Less |
Product Title
Select an issue
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?