Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bulwark™ FR Women's Lightweight Flame Resistant Cargo Pant

Catalog No. 17200802 Shop All Red Kap Industries Products
Change view
Click to view available options
Size:
4
6
8
10
12
14
16
18
20
22
24
Size (English) Inseam:
28 in.
30 in.
32 in.
34 in.
44 product options available for selection
Product selection table with 44 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Size (English) Inseam
17-200-802 20 34 in.
17-200-803 22 28 in.
17-200-804 22 30 in.
17-200-808 24 30 in.
17-200-806 22 34 in.
17-200-807 24 28 in.
17-200-805 22 32 in.
17-200-809 24 32 in.
17-200-810 24 34 in.
17-150-465 4 28 in.
17-150-466 4 30 in.
17-150-470 6 30 in.
17-150-469 6 28 in.
17-150-467 4 32 in.
17-150-474 8 30 in.
17-150-472 6 34 in.
17-150-471 6 32 in.
17-150-473 8 28 in.
17-150-468 4 34 in.
17-150-475 8 32 in.
17-150-476 8 34 in.
17-150-478 10 30 in.
17-150-481 12 28 in.
17-150-477 10 28 in.
17-150-480 10 34 in.
17-150-482 12 30 in.
17-150-479 10 32 in.
17-150-483 12 32 in.
17-150-487 14 32 in.
17-150-485 14 28 in.
17-150-486 14 30 in.
17-150-484 12 34 in.
17-150-488 14 34 in.
17-150-489 16 28 in.
17-150-490 16 30 in.
17-150-491 16 32 in.
17-150-493 18 28 in.
17-150-492 16 34 in.
17-150-494 18 30 in.
17-150-497 20 28 in.
17-150-496 18 34 in.
17-150-495 18 32 in.
17-150-498 20 30 in.
17-150-499 20 32 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 44 options available.
44 options
This item is not returnable. View return policy
Catalog No. 17-200-802 Supplier Bulwark™ FR Supplier No. PMU3NV 20 34U
Add to Cart
Add to Cart
This item is not returnable. View return policy

If storage and breathability are what you're looking for, then this Lightweight CAT 2 AR/FR Cargo Pant is the right FR pant for you. Seven total pockets, including a cargo pocket on each leg ensures you've got plenty of room to store all your tools.

  • Primary Closure: Hook and Eye
  • Wash Care: Industrial Laundry - Light Soil, Home Wash
  • Fabric: Flame resistant, 7 oz. (235 g/m2)
  • Blend: 47% Modacrylic/37% Lyocell/16% Aramid
  • Closure: Waistband with hook-and-bar closure
  • Pocket: Two front slack-style pockets and two cargo utility pockets, Left cargo pocket has two snap closures and right utility pocket is open
  • Features:
    • Waistband with hook-and-bar closure
    • Two utility pockets
    • Left cargo pocket has two snap closures
    • Right utility pocket is open
  • Product Collection: Cooltouch 2
  • Protection: ATPV Arc Rating: 11
  • Country of Origin: Imported, Made in USA Fabric
  • Fit Details: Sits at the waist. Regular in the seat and thigh. Slightly tapered leg.
  • Gender: Female
  • Materials: Modacrylic, Lyocell, Aramid
  • Brand: Bulwark™

Specifications

Size 20
Product Type Flame-resistant Cargo Pants
Gender Women's
Material Modacrylic, Lyocell, Polyamide-Imide, Aramid
Color Navy
No. of Pockets 5
Closure Type Button
Resistant To Flame
Hazard Risk Category HRC2
Arc Flash Rating ATPV 10.1 Calories/cm2
Certifications/Compliance Meets NFPA 2112
Size (English) Inseam 34 in.
Style Unhemmed
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.