Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bulwark™ FR Women's Lightweight Flame Resistant Work Pant

Catalog No. 17150532 Shop All Red Kap Industries Products
Change view
Click to view available options
Size:
4
6
8
10
12
14
16
18
20
22
24
Size (English) Inseam:
28 in.
30 in.
32 in.
34 in.
44 product options available for selection
Product selection table with 44 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Size (English) Inseam
17-150-532 8 32 in.
17-150-522 4 28 in.
17-150-523 4 30 in.
17-150-524 4 32 in.
17-150-525 4 34 in.
17-150-526 6 28 in.
17-150-529 6 34 in.
17-150-527 6 30 in.
17-150-528 6 32 in.
17-150-531 8 30 in.
17-150-530 8 28 in.
17-150-534 10 28 in.
17-150-533 8 34 in.
17-150-538 12 28 in.
17-150-535 10 30 in.
17-150-536 10 32 in.
17-150-537 10 34 in.
17-150-539 12 30 in.
17-150-540 12 32 in.
17-150-542 14 28 in.
17-150-541 12 34 in.
17-150-543 14 30 in.
17-150-545 14 34 in.
17-150-546 16 28 in.
17-150-544 14 32 in.
17-150-547 16 30 in.
17-150-548 16 32 in.
17-150-549 16 34 in.
17-150-553 18 34 in.
17-150-555 20 30 in.
17-150-550 18 28 in.
17-150-551 18 30 in.
17-150-552 18 32 in.
17-150-554 20 28 in.
17-150-556 20 32 in.
17-150-557 20 34 in.
17-150-558 22 28 in.
17-150-563 24 30 in.
17-150-560 22 32 in.
17-150-561 22 34 in.
17-150-559 22 30 in.
17-150-562 24 28 in.
17-150-564 24 32 in.
17-150-565 24 34 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 44 options available.
44 options
This item is not returnable. View return policy
Catalog No. 17-150-532 Supplier Bulwark™ FR Supplier No. PMW3NV 08 32
Add to Cart
Add to Cart
This item is not returnable. View return policy

This women's work pant provides a stylish design with an interlined waistband with button closure, two slack style front pockets, two set-in hip pockets and a left hip pocket with button closure.

  • Primary Closure: Button
  • Wash Care: Industrial Laundry - Heavy Soil, Industrial Laundry - Light Soil, Home Wash
  • Fabric: Flame resistant, 7 oz. (235 g/m2)
  • Blend: 47% Modacryclic/37%Lyocell/16% Aramid
  • Closure: Interlined waistband with button closure
  • Pocket: Two slack style front pockets, two set-in hip pockets and left hip pocket with button closure
  • Features:
    • Interlined waistband with button closure
    • Two slack style front pockets
    • Two set-in hip pockets
    • Left hip pocket with button closure
  • Product Collection: Cooltouch 2
  • Protection: ATPV Arc Rating: 11
  • Country of Origin: Imported, Made in USA Fabric
  • Fit Details: Sits at the waist. Regular in the seat and thigh. Slightly tapered leg.
  • Gender: Female
  • Materials: Modacrylic, Lyocell, Aramid
  • Brand: Bulwark™

Specifications

Size 8
Gender Women's
Product Type Flame-resistant Work Pants
Material Modacrylic, Lyocell, Polyamide-Imide, Aramid
Color Navy
No. of Pockets 4
Closure Type Button
Hazard Risk Category HRC2
Arc Flash Rating ATPV 10.1 Calories/cm2
Certifications/Compliance Meets NFPA 2112, UL
Size (English) Inseam 32 in.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.