Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bulwark™ FR Men's Relaxed Excel FR™ Flame Resistant Jean, Dark Denim

Catalog No. 19296606 Shop All Red Kap Industries Products
Change view
Click to view available options
Size:
28 x 30 in.
28 x 32 in.
28 x 34 in.
30 x 30 in.
30 x 32 in.
30 x 34 in.
32 x 30 in.
32 x 32 in.
32 x 34 in.
34 x 30 in.
34 x 32 in.
34 x 34 in.
36 x 30 in.
36 x 32 in.
36 x 34 in.
38 x 30 in.
38 x 32 in.
38 x 34 in.
40 x 30 in.
40 x 32 in.
40 x 34 in.
42 x 30 in.
42 x 32 in.
42 x 34 in.
44 x 30 in.
44 x 32 in.
44 x 34 in.
46 x 30 in.
46 x 32 in.
46 x 34 in.
48 x 30 in.
48 x 32 in.
48 x 34 in.
50 x 30 in.
50 x 32 in.
50 x 34 in.
54 x 30 in.
37 product options available for selection
Product selection table with 37 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-296-606 38 x 30 in.
19-296-591 28 x 30 in.
19-296-592 28 x 32 in.
19-296-593 28 x 34 in.
19-296-594 30 x 30 in.
19-296-596 30 x 34 in.
19-296-597 32 x 30 in.
19-296-595 30 x 32 in.
19-296-598 32 x 32 in.
19-296-599 32 x 34 in.
19-296-600 34 x 30 in.
19-296-602 34 x 34 in.
19-296-601 34 x 32 in.
19-296-603 36 x 30 in.
19-296-604 36 x 32 in.
19-296-605 36 x 34 in.
19-296-607 38 x 32 in.
19-296-608 38 x 34 in.
19-296-609 40 x 30 in.
19-296-610 40 x 32 in.
19-296-611 40 x 34 in.
19-296-613 42 x 32 in.
19-296-612 42 x 30 in.
19-296-614 42 x 34 in.
19-296-615 44 x 30 in.
19-296-620 46 x 34 in.
19-296-616 44 x 32 in.
19-296-617 44 x 34 in.
19-296-619 46 x 32 in.
19-296-621 48 x 30 in.
19-296-618 46 x 30 in.
19-296-622 48 x 32 in.
19-296-626 50 x 34 in.
19-296-624 50 x 30 in.
19-296-627 54 x 30 in.
19-296-623 48 x 34 in.
19-296-625 50 x 32 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 37 options available.
37 options
This item is not returnable. View return policy
Catalog No. 19-296-606 Supplier Bulwark™ FR Supplier No. PEJ2DD 38 30
Add to Cart
Add to Cart
This item is not returnable. View return policy

This CAT 2 AR/FR Denim Jean is NFPA™ 2112 compliant. It provides a relaxed fit with classic five-pocket jean styling for improved comfort with FR protection.

  • Primary Closure: Zipper
  • Wash Care: Industrial Laundry - Heavy Soil, Industrial Laundry - Light Soil, Home Wash
  • Fabric: EXCEL FR™ Flame resistant, 12.5 oz. (425 g/m2)
  • Blend: 100% Cotton Denim
  • Closure: One-piece waistband with button closure
  • Pocket: Five jean-style pockets
  • Features:
    • Sits at natural waist
    • Relaxed seat and thigh with straight leg opening
    • One-piece waistband with button closure
  • Product Collection: Excel FR, Denim
  • Protection: ATPV Arc Rating: 18
  • Country of Origin: Imported, Made in USA Fabric
  • Fit Details: Sits just below the waist. Relaxed in the seat and thigh. Straight leg.
  • Gender: Male
  • Materials: Cotton
  • Brand: Bulwark™

Specifications

Size 38 x 30 in.
Product Type Flame-resistant Jeans
Gender Men's
Material Denim
Color Blue
No. of Pockets 5
Closure Type Button
Resistant To Flame
Hazard Risk Category HRC2
Arc Flash Rating ATPV 18 Calories/cm2
Size (English) Inseam 30 in.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.