Description
- Banded, topstitched, button-down collar
- Two chest pockets with button flap closures and sewn-in pencil stall
- Placket front with button closure
- Topstitched cuff with button closure
- Tailored sleeve plackets
- Home wash and industrial wash
- Protection: Arc Rating ATPV 8.6 cal./cm2
- Available in navy and light blue colors
Specifications
Specifications
| Size | Large (Long) |
| Color | Navy |
| Hazard Risk Category | Flame-resistant |