Learn More
Bulwark™ FR Excel FR™ Comfortouch™ 7 oz. Dress Uniform Shirts
Catalog No. 19295119 Shop All Red Kap Industries Products
Change view
Click to view available options
Color:
Light Blue
Navy
Size:
2X-Large (Long)
2X-Large (Regular)
3X-Large (Regular)
Large (Long)
Large (Regular)
Medium (Long)
Medium (Regular)
Small (Regular)
X-Large (Long)
X-Large (Regular)
17 product options available for selection
Product selection table with 17 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
19-295-119 Light Blue 3X-Large (Regular)
18-120-270 Navy Medium (Regular)
18-120-269 Navy Small (Regular)
18-120-271 Navy Medium (Long)
18-120-273 Navy Large (Long)
18-120-272 Navy Large (Regular)
18-120-275 Navy X-Large (Long)
18-120-276 Navy 2X-Large (Regular)
18-120-278 Navy 3X-Large (Regular)
18-120-277 Navy 2X-Large (Long)
19-295-114 Light Blue Small (Regular)
19-295-115 Light Blue Medium (Regular)
19-295-116 Light Blue Large (Regular)
19-295-117 Light Blue X-Large (Regular)
19-295-118 Light Blue 2X-Large (Regular)
19-295-121 Light Blue Large (Long)
19-295-120 Light Blue Medium (Long)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 17 options available.
17 options
This item is not returnable. View return policy
Catalog No. 19-295-119 Supplier Bulwark™ FR Supplier No. SLU2LB RG 3XL
Add to Cart
Add to Cart
This item is not returnable. View return policy
Designed with flame-resistant fabric, 7 oz. (235g/m2) 88% Cotton/12% Nylon
  • Banded, topstitched, button-down collar
  • Two chest pockets with button flap closures and sewn-in pencil stall
  • Placket front with button closure
  • Topstitched cuff with button closure
  • Tailored sleeve plackets
  • Home wash and industrial wash
  • Protection: Arc Rating ATPV 8.6 cal./cm2
  • Available in navy and light blue colors
Specifications
Size 3X-Large (Regular)
Color Light Blue
Hazard Risk Category Flame-resistant