Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bulwark™ FR Men's Heavyweight Excel FR™ Flame Resistant Denim Dungaree

Catalog No. 17150303 Shop All Red Kap Industries Products
Change view
Click to view available options
Size:
28
30
32
34
36
38
40
42
44
46
48
50
Size (English) Inseam:
30 in.
32 in.
34 in.
37 in.
48 product options available for selection
Product selection table with 48 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Size (English) Inseam
17-150-303 30 37 in.
17-200-798 28 30 in.
17-200-801 28 37 in.
17-200-799 28 32 in.
17-200-800 28 34 in.
17-150-300 30 30 in.
17-150-301 30 32 in.
17-150-302 30 34 in.
17-150-304 32 30 in.
17-150-305 32 32 in.
17-150-307 32 37 in.
17-150-310 34 34 in.
17-150-311 34 37 in.
17-150-308 34 30 in.
17-150-309 34 32 in.
17-150-306 32 34 in.
17-150-312 36 30 in.
17-150-313 36 32 in.
17-150-316 38 30 in.
17-150-315 36 37 in.
17-150-314 36 34 in.
17-150-317 38 32 in.
17-150-319 38 37 in.
17-150-318 38 34 in.
17-150-320 40 30 in.
17-150-321 40 32 in.
17-150-322 40 34 in.
17-150-323 40 37 in.
17-150-324 42 30 in.
17-150-327 42 37 in.
17-150-325 42 32 in.
17-150-329 44 32 in.
17-150-328 44 30 in.
17-150-326 42 34 in.
17-150-331 44 37 in.
17-150-330 44 34 in.
17-150-333 46 32 in.
17-150-332 46 30 in.
17-150-334 46 34 in.
17-150-336 48 30 in.
17-150-337 48 32 in.
17-150-335 46 37 in.
17-150-338 48 34 in.
17-150-339 48 37 in.
17-150-340 50 30 in.
17-150-341 50 32 in.
17-150-342 50 34 in.
17-150-343 50 37 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 48 options available.
48 options
This item is not returnable. View return policy
Catalog No. 17-150-303 Supplier Bulwark™ FR Supplier No. PEJ8DW 30 37U
Add to Cart
Add to Cart
This item is not returnable. View return policy

This Heavyweight CAT 2 AR/FR Dungaree Style Denim jean is NFPA™ 2112 compliant. Convenient features like a hammer loop on left leg and utility loop on waistband ensure you've got a dedicated place for all your tools. Loose fit improves comfort and a wider leg opening fits easily over boots.

  • Primary Closure: Zipper
  • Wash Care: Industrial Laundry - Heavy Soil, Industrial Laundry - Light Soil, Home Wash
  • Fabric: Excel FR™ Flame resistant, 14.75 oz. (500 g/m2)
  • Blend: 100% Cotton Denim
  • Closure: Zipper fly with single metal button on waistband
  • Pocket: Two deep scoop pockets,Two oversized reinforced hip pockets,One rule pocket on right leg with wrap around utility pocket
  • Features:
    • Sits at natural waist
    • Loose seat and thigh with wider leg opening
    • Two deep scoop pockets and two oversized reinforced hip pockets with rule pocket on right leg and wrap around utility pocket
    • Soft pre-wash for comfort and look
  • Product Collection: Excel FR, Denim
  • Protection: ATPV Arc Rating: 21
  • Country of Origin: Imported, Made in USA Fabric
  • Fit Details: Sits at the waist. Extra room in the seat and thigh. Relaxed in the leg.
  • Gender: Male
  • Materials: Cotton
  • Brand: Bulwark™

Specifications

Size 30
Gender Men's
Product Type Flame-resistant Dungarees
Material Cotton
Color Blue
No. of Pockets 5
Closure Type Concealed button closure
Finish Ammonia Cure
Resistant To Flame
Hazard Risk Category HRC2
Arc Flash Rating ATPV 18.0 Calories/cm2
Certifications/Compliance Meets NFPA 2112, UL, and 70E compliant
Size (English) Inseam 37 in.
Style Unhemmed
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.