Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
- Home
- Products
- Personal Protective Equipment
- Safety Clothing
- Pants, Trousers, and Leggings
- Heat and Fire Resistant Pants, Trousers, and Leggings
- Bulwark™ FR Men's Heavyweight Excel FR™ Flame Resistant Denim Dungaree
Bulwark™ FR Men's Heavyweight Excel FR™ Flame Resistant Denim Dungaree
Catalog No. 17150332 Shop All Red Kap Industries Products
Change view
Click to view available options
Size:
28
30
32
34
36
38
40
42
44
46
48
50
Size (English) Inseam:
30 in.
32 in.
34 in.
37 in.
48 product options available for selection
| Catalog No. | Size | Size (English) Inseam |
|---|---|---|
| 17-150-332 | 46 | 30 in. |
| 17-200-798 | 28 | 30 in. |
| 17-200-801 | 28 | 37 in. |
| 17-200-799 | 28 | 32 in. |
| 17-200-800 | 28 | 34 in. |
| 17-150-300 | 30 | 30 in. |
| 17-150-301 | 30 | 32 in. |
| 17-150-302 | 30 | 34 in. |
| 17-150-303 | 30 | 37 in. |
| 17-150-304 | 32 | 30 in. |
| 17-150-305 | 32 | 32 in. |
| 17-150-307 | 32 | 37 in. |
| 17-150-310 | 34 | 34 in. |
| 17-150-311 | 34 | 37 in. |
| 17-150-308 | 34 | 30 in. |
| 17-150-309 | 34 | 32 in. |
| 17-150-306 | 32 | 34 in. |
| 17-150-312 | 36 | 30 in. |
| 17-150-313 | 36 | 32 in. |
| 17-150-316 | 38 | 30 in. |
| 17-150-315 | 36 | 37 in. |
| 17-150-314 | 36 | 34 in. |
| 17-150-317 | 38 | 32 in. |
| 17-150-319 | 38 | 37 in. |
| 17-150-318 | 38 | 34 in. |
| 17-150-320 | 40 | 30 in. |
| 17-150-321 | 40 | 32 in. |
| 17-150-322 | 40 | 34 in. |
| 17-150-323 | 40 | 37 in. |
| 17-150-324 | 42 | 30 in. |
| 17-150-327 | 42 | 37 in. |
| 17-150-325 | 42 | 32 in. |
| 17-150-329 | 44 | 32 in. |
| 17-150-328 | 44 | 30 in. |
| 17-150-326 | 42 | 34 in. |
| 17-150-331 | 44 | 37 in. |
| 17-150-330 | 44 | 34 in. |
| 17-150-333 | 46 | 32 in. |
| 17-150-334 | 46 | 34 in. |
| 17-150-336 | 48 | 30 in. |
| 17-150-337 | 48 | 32 in. |
| 17-150-335 | 46 | 37 in. |
| 17-150-338 | 48 | 34 in. |
| 17-150-339 | 48 | 37 in. |
| 17-150-340 | 50 | 30 in. |
| 17-150-341 | 50 | 32 in. |
| 17-150-342 | 50 | 34 in. |
| 17-150-343 | 50 | 37 in. |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 48 options available.
This item is not returnable.
View return policy
This item is not returnable.
View return policy
Description
- Primary Closure: Zipper
- Wash Care: Industrial Laundry - Heavy Soil, Industrial Laundry - Light Soil, Home Wash
- Fabric: Excel FR™ Flame resistant, 14.75 oz. (500 g/m2)
- Blend: 100% Cotton Denim
- Closure: Zipper fly with single metal button on waistband
- Pocket: Two deep scoop pockets,Two oversized reinforced hip pockets,One rule pocket on right leg with wrap around utility pocket
- Features:
- Sits at natural waist
- Loose seat and thigh with wider leg opening
- Two deep scoop pockets and two oversized reinforced hip pockets with rule pocket on right leg and wrap around utility pocket
- Soft pre-wash for comfort and look
- Product Collection: Excel FR, Denim
- Protection: ATPV Arc Rating: 21
- Country of Origin: Imported, Made in USA Fabric
- Fit Details: Sits at the waist. Extra room in the seat and thigh. Relaxed in the leg.
- Gender: Male
- Materials: Cotton
- Brand: Bulwark™
Specifications
Specifications
| Size | 46 |
| Gender | Men's |
| Product Type | Flame-resistant Dungarees |
| Material | Cotton |
| Color | Blue |
| No. of Pockets | 5 |
| Closure Type | Concealed button closure |
| Finish | Ammonia Cure |
| Resistant To | Flame |
| Hazard Risk Category | HRC2 |
| Arc Flash Rating | ATPV 18.0 Calories/cm2 |
| Certifications/Compliance | Meets NFPA 2112, UL, and 70E compliant |
| Size (English) Inseam | 30 in. |
| Show More Show Less |
Product Title
Select an issue
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?