Learn More
Bulwark™ FR Men's Midweight Excel FR™ Flame Resistant Khaki Work Pant, Waist Sizes 28 to 40 in.
Shop All Red Kap Industries ProductsDescription
- Primary Closure: Button
- Wash Care: Industrial Laundry - Heavy Soil, Industrial Laundry - Light Soil, Home Wash
- Fabric: Excel FR™ Flame resistant, 9 oz. (305 g/m2) Twill
- Blend: 100% Cotton
- Closure: Waistband with button closure
- Pocket: Two slack-style front pockets and left hip pocket with button closure
- Features:
- Waistband with button closure
- Two slack-style pockets
- Left hip pocket with button closure
- Product Collection: Excel FR
- Protection: ATPV Arc Rating: 11
- Country of Origin: Imported, Made in USA Fabric
- Fit Details: Sits at the waist. Regular in the seat and thigh. Slightly tapered leg.
- Gender: Male
- Materials: Cotton
- Brand: Bulwark™
Specifications
Specifications
| Size | 34 |
| Product Type | Flame-resistant Work Pants |
| Gender | Men's |
| Material | Cotton |
| Color | Khaki |
| No. of Pockets | 4 |
| Closure Type | Button |
| Resistant To | Flame |
| Hazard Risk Category | HRC2 |
| Certifications/Compliance | Meets NFPA 2112 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.