Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bulwark™ FR Men's Midweight Excel FR™ Flame Resistant Khaki Work Pant, Waist Sizes 28 to 40 in.

Catalog No. 17150375 Shop All Red Kap Industries Products
Change view
Click to view available options
Size:
28
29
30
31
32
33
34
36
38
40
Size (English) Inseam:
30 in.
32 in.
34 in.
36 in.
37 in.
40 product options available for selection
Product selection table with 40 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Size (English) Inseam
17-150-375 36 37 in.
17-150-344 28 30 in.
17-150-348 29 30 in.
17-150-349 29 32 in.
17-150-346 28 34 in.
17-150-345 28 32 in.
17-150-347 28 36 in.
17-150-350 29 34 in.
17-150-351 29 36 in.
17-150-354 30 34 in.
17-150-356 31 30 in.
17-150-353 30 32 in.
17-150-355 30 37 in.
17-150-352 30 30 in.
17-150-358 31 34 in.
17-150-357 31 32 in.
17-150-359 31 37 in.
17-150-362 32 34 in.
17-150-360 32 30 in.
17-150-363 32 37 in.
17-150-366 33 34 in.
17-150-365 33 32 in.
17-150-361 32 32 in.
17-150-364 33 30 in.
17-150-367 33 37 in.
17-150-369 34 32 in.
17-150-371 34 37 in.
17-150-370 34 34 in.
17-150-368 34 30 in.
17-150-373 36 32 in.
17-150-372 36 30 in.
17-150-374 36 34 in.
17-150-377 38 32 in.
17-150-381 40 32 in.
17-150-380 40 30 in.
17-150-379 38 37 in.
17-150-378 38 34 in.
17-150-376 38 30 in.
17-150-382 40 34 in.
17-150-383 40 36 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 40 options available.
40 options
This item is not returnable. View return policy
Catalog No. 17-150-375 Supplier Bulwark™ FR Supplier No. PEW2KH 36 37U
Add to Cart
Add to Cart
This item is not returnable. View return policy

This midweight work pant offers a classic look with CAT 2 AR/FR protection and NFPA™ 2112 compliance.

  • Primary Closure: Button
  • Wash Care: Industrial Laundry - Heavy Soil, Industrial Laundry - Light Soil, Home Wash
  • Fabric: Excel FR™ Flame resistant, 9 oz. (305 g/m2) Twill
  • Blend: 100% Cotton
  • Closure: Waistband with button closure
  • Pocket: Two slack-style front pockets and left hip pocket with button closure
  • Features:
    • Waistband with button closure
    • Two slack-style pockets
    • Left hip pocket with button closure
  • Product Collection: Excel FR
  • Protection: ATPV Arc Rating: 11
  • Country of Origin: Imported, Made in USA Fabric
  • Fit Details: Sits at the waist. Regular in the seat and thigh. Slightly tapered leg.
  • Gender: Male
  • Materials: Cotton
  • Brand: Bulwark™

Specifications

Size 36
Gender Men's
Product Type Flame-resistant Work Pants
Material Cotton
Color Khaki
No. of Pockets 4
Closure Type Button
Resistant To Flame
Hazard Risk Category HRC2
Certifications/Compliance Meets NFPA 2112
Size (English) Inseam 37 in.
Style Unhemmed
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.