Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bulwark™ FR Men's Midweight Excel FR™ Flame Resistant Khaki Work Pant, Waist Sizes 42 to 50 in.

Catalog No. 17150386 Shop All Red Kap Industries Products
Change view
Click to view available options
Size:
42
44
46
48
50
Size (English) Inseam:
30 in.
32 in.
34 in.
36 in.
20 product options available for selection
Product selection table with 20 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Size (English) Inseam
17-150-386 42 34 in.
17-150-391 44 36 in.
17-150-388 44 30 in.
17-150-384 42 30 in.
17-150-389 44 32 in.
17-150-385 42 32 in.
17-150-390 44 34 in.
17-150-387 42 36 in.
17-150-392 46 30 in.
17-150-396 48 30 in.
17-150-397 48 32 in.
17-150-394 46 34 in.
17-150-398 48 34 in.
17-150-399 48 36 in.
17-150-393 46 32 in.
17-150-395 46 36 in.
17-150-400 50 30 in.
17-150-402 50 34 in.
17-150-403 50 36 in.
17-150-401 50 32 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 20 options available.
20 options
This item is not returnable. View return policy
Catalog No. 17-150-386 Supplier Bulwark™ FR Supplier No. PEW2KH 42 34
Add to Cart
Add to Cart
This item is not returnable. View return policy

This midweight work pant offers a classic look with CAT 2 AR/FR protection and NFPA™ 2112 compliance.

  • Primary Closure: Button
  • Wash Care: Industrial Laundry - Heavy Soil, Industrial Laundry - Light Soil, Home Wash
  • Fabric: Excel FR™ Flame resistant, 9 oz. (305 g/m2) Twill
  • Blend: 100% Cotton
  • Closure: Waistband with button closure
  • Pocket: Two slack-style front pockets and left hip pocket with button closure
  • Features:
    • Waistband with button closure
    • Two slack-style pockets
    • Left hip pocket with button closure
  • Product Collection: Excel FR
  • Protection: ATPV Arc Rating: 11
  • Country of Origin: Imported, Made in USA Fabric
  • Fit Details: Sits at the waist. Regular in the seat and thigh. Slightly tapered leg.
  • Gender: Male
  • Materials: Cotton
  • Brand: Bulwark™

Specifications

Size 42
Product Type Flame-resistant Work Pants
Gender Men's
Material Cotton
Color Khaki
No. of Pockets 4
Closure Type Button
Finish Ammonia Cure
Resistant To Flame
Hazard Risk Category HRC2
Arc Flash Rating ATPV 10.6 Calories/cm2
Certifications/Compliance Meets NFPA 2112
Size (English) Inseam 34 in.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.