Description
- Three button placket
- Knit henley collar
- Rib-knit cuff
- Single spade chest pocket
- Tagless brand label and warning label
- Home wash
- Protection: Arc Rating ATPV 9.6 cal./cm2
Specifications
Specifications
| Size | Small (Regular) |
| Color | Navy |
| Neck Style | Henley |
| Arc Flash Rating | ATPV 9.6 cal./cm2 |