Learn More
Bulwark™ FR Excel FR™ Long Sleeve Tagless Henley Men's Shirts
Catalog No. 18120307 Shop All Red Kap Industries Products
Change view
Click to view available options
Color:
Gray
Navy
Size:
2X-Large (Long)
2X-Large (Regular)
3X-Large (Regular)
4X-Large (Regular)
Large (Long)
Large (Regular)
Medium (Regular)
Small (Regular)
X-Large (Long)
X-Large (Regular)
19 product options available for selection
Product selection table with 19 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
18-120-307 Navy Medium (Regular)
18-120-306 Navy Small (Regular)
18-120-308 Navy Large (Regular)
18-120-309 Navy Large (Long)
18-120-310 Navy X-Large (Regular)
18-120-314 Navy 3X-Large (Regular)
18-120-311 Navy X-Large (Long)
18-120-312 Navy 2X-Large (Regular)
18-120-313 Navy 2X-Large (Long)
18-120-315 Navy 4X-Large (Regular)
18-120-317 Gray Medium (Regular)
18-120-316 Gray Small (Regular)
18-120-318 Gray Large (Regular)
18-120-321 Gray X-Large (Long)
18-120-319 Gray Large (Long)
18-120-322 Gray 2X-Large (Regular)
18-120-324 Gray 3X-Large (Regular)
18-120-325 Gray 4X-Large (Regular)
18-120-323 Gray 2X-Large (Long)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 19 options available.
19 options
This item is not returnable. View return policy
Catalog No. 18-120-307 Supplier Bulwark™ FR Supplier No. SEL2NV RG M
Add to Cart
Add to Cart
This item is not returnable. View return policy
Designed with flame-resistant fabric, 210g/m2 (6.25 oz.) Interlock FR 100% Cotton
  • Three button placket
  • Knit henley collar
  • Rib-knit cuff
  • Single spade chest pocket
  • Tagless brand label and warning label
  • Home wash
  • Protection: Arc Rating ATPV 9.6 cal./cm2
Specifications
Size Medium (Regular)
Color Navy
Neck Style Henley
Arc Flash Rating ATPV 9.6 cal./cm2