Learn More
Bulwark™ FR Men's Classic Heavyweight Excel FR™ Flame Resistant Jean, Blue Denim
Shop All Red Kap Industries ProductsDescription
- Primary Closure: Button
- Wash Care: Industrial Laundry - Heavy Soil, Industrial Laundry - Light Soil, Home Wash
- Fabric: EXCEL FR™ Flame resistant, 14.75 oz. (500 g/m2)
- Blend: 100% Pre-Washed Cotton Denim
- Closure: Button closure
- Pocket: Five jean-style pockets
- Features:
- Sits at natural waist
- Relaxed seat and thigh
- Soft pre-wash for comfort and look
- Two-needle felled seam construction
- Product Collection: Excel FR, Denim
- Protection: ATPV Arc Rating: 21
- Country of Origin: Imported, Made in USA Fabric
- Fit Details: Sits at waist. Regular in seat and thigh. Slightly tapered leg.
- Gender: Male
- Materials: Cotton
- Brand: Bulwark™
Specifications
Specifications
| Size | 36 x 32 in. |
| Gender | Men's |
| Product Type | Flame-resistant Jeans |
| Material | Cotton |
| Color | Blue |
| No. of Pockets | 5 |
| Closure Type | Button |
| Hazard Risk Category | HRC2 |
| Certifications/Compliance | Meets NFPA 2112 |
| Size (English) Inseam | 32 in. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.