Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bulwark™ FR Men's Classic Heavyweight Excel FR™ Flame Resistant Jean, Blue Denim

Catalog No. 17200795 Shop All Red Kap Industries Products
Change view
Click to view available options
Size:
28 x 30 in.
28 x 32 in.
28 x 34 in.
30 x 30 in.
30 x 32 in.
30 x 34 in.
31 x 30 in.
31 x 32 in.
31 x 34 in.
32 x 30 in.
32 x 32 in.
32 x 34 in.
33 x 30 in.
33 x 32 in.
33 x 34 in.
34 x 30 in.
34 x 32 in.
34 x 34 in.
35 x 30 in.
35 x 32 in.
35 x 34 in.
36 x 30 in.
36 x 32 in.
36 x 34 in.
38 x 30 in.
38 x 32 in.
38 x 34 in.
40 x 30 in.
40 x 32 in.
40 x 34 in.
42 x 30 in.
42 x 32 in.
42 x 34 in.
44 x 30 in.
44 x 32 in.
44 x 34 in.
46 x 30 in.
46 x 32 in.
46 x 34 in.
48 x 30 in.
48 x 32 in.
48 x 34 in.
50 x 30 in.
50 x 32 in.
50 x 34 in.
45 product options available for selection
Product selection table with 45 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-200-795 50 x 30 in.
17-200-755 28 x 34 in.
17-200-753 28 x 30 in.
17-200-754 28 x 32 in.
17-200-756 30 x 30 in.
17-200-759 31 x 30 in.
17-200-760 31 x 32 in.
17-200-758 30 x 34 in.
17-200-757 30 x 32 in.
17-200-761 31 x 34 in.
17-200-764 32 x 34 in.
17-200-763 32 x 32 in.
17-200-762 32 x 30 in.
17-200-765 33 x 30 in.
17-200-766 33 x 32 in.
17-200-767 33 x 34 in.
17-200-768 34 x 30 in.
17-200-769 34 x 32 in.
17-200-773 35 x 34 in.
17-200-770 34 x 34 in.
17-200-771 35 x 30 in.
17-200-772 35 x 32 in.
17-200-775 36 x 32 in.
17-200-774 36 x 30 in.
17-200-776 36 x 34 in.
17-200-777 38 x 30 in.
17-200-778 38 x 32 in.
17-200-779 38 x 34 in.
17-200-782 40 x 34 in.
17-200-780 40 x 30 in.
17-200-785 42 x 34 in.
17-200-783 42 x 30 in.
17-200-784 42 x 32 in.
17-200-781 40 x 32 in.
17-200-786 44 x 30 in.
17-200-787 44 x 32 in.
17-200-793 48 x 32 in.
17-200-788 44 x 34 in.
17-200-789 46 x 30 in.
17-200-791 46 x 34 in.
17-200-794 48 x 34 in.
17-200-790 46 x 32 in.
17-200-792 48 x 30 in.
17-200-796 50 x 32 in.
17-200-797 50 x 34 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 45 options available.
45 missing translation for 'options'
This item is not returnable. View return policy
Catalog No. 17-200-795 missing translation for 'mfr' Bulwark™ FR missing translation for 'supplierNo' PEJ4DW 50 30
Add to Cart
Add to Cart
This item is not returnable. View return policy

This Heavyweight CAT 2 AR/FR Denim Jean is NFPA™ 2112 compliant. It offers a classic workwear look and fit with FR protection and five-pocket jean styling. Classic fit sits at the natural waist and has a slimmer leg opening that will fit over boots.

  • Primary Closure: Button
  • Wash Care: Industrial Laundry - Heavy Soil, Industrial Laundry - Light Soil, Home Wash
  • Fabric: EXCEL FR™ Flame resistant, 14.75 oz. (500 g/m2)
  • Blend: 100% Pre-Washed Cotton Denim
  • Closure: Button closure
  • Pocket: Five jean-style pockets
  • Features:
    • Sits at natural waist
    • Relaxed seat and thigh
    • Soft pre-wash for comfort and look
    • Two-needle felled seam construction
  • Product Collection: Excel FR, Denim
  • Protection: ATPV Arc Rating: 21
  • Country of Origin: Imported, Made in USA Fabric
  • Fit Details: Sits at waist. Regular in seat and thigh. Slightly tapered leg.
  • Gender: Male
  • Materials: Cotton
  • Brand: Bulwark™

Specifications

Size 50 x 30 in.
Gender Men's
Product Type Flame-resistant Jeans
Material Cotton
Color Blue
No. of Pockets 5
Closure Type Button
Hazard Risk Category HRC2
Arc Flash Rating ATPV 20.7 Calories/cm2
Certifications/Compliance Meets NFPA 2112
Size (English) Inseam 30 in.
Size (English) Waist 50
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.