Learn More
Bulwark™ FR Men's Relaxed Excel FR™ Flame Resistant Jean, Dark Denim
Shop All Red Kap Industries ProductsDescription
- Primary Closure: Zipper
- Wash Care: Industrial Laundry - Heavy Soil, Industrial Laundry - Light Soil, Home Wash
- Fabric: EXCEL FR™ Flame resistant, 12.5 oz. (425 g/m2)
- Blend: 100% Cotton Denim
- Closure: One-piece waistband with button closure
- Pocket: Five jean-style pockets
- Features:
- Sits at natural waist
- Relaxed seat and thigh with straight leg opening
- One-piece waistband with button closure
- Product Collection: Excel FR, Denim
- Protection: ATPV Arc Rating: 18
- Country of Origin: Imported, Made in USA Fabric
- Fit Details: Sits just below the waist. Relaxed in the seat and thigh. Straight leg.
- Gender: Male
- Materials: Cotton
- Brand: Bulwark™
Specifications
Specifications
| Size | 36 |
| Gender | Men's |
| Product Type | Flame-resistant Jeans |
| Material | Cotton |
| Color | Blue |
| No. of Pockets | 5 |
| Closure Type | Button |
| Finish | Ammonia Cure |
| Resistant To | Flame |
| Hazard Risk Category | HRC2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.