Learn More
Description
- Primary Closure: No Closure
- Wash Care: Home Wash
- Fabric: Excel FR™ Flame resistant, 5.5 oz. (185 g/m2)
- Blend: 90% Cotton/10% Polyester
- Finish: Moisture management
- Collar: Self-fabric crewneck collar
- Features:
- Self-fabric crewneck collar
- Hemmed, raglan sleeve
- Contrast mesh full underarm gussets
- Heat seal Bulwark™ triangle centered at neck to show wearer has on FR undergarment
- Product Collection: Excel FR
- Protection: ATPV Arc Rating: 6.4
- Country of Origin: Imported, Made in USA Fabric
- Fit Details: Slim Fit
- Gender: Male
- Materials: Cotton/Polyester
- Brand: Bulwark™
Specifications
Specifications
| Size | Small |
| Gender | Men's |
| Material | Cotton, Polyester |
| Color | Gray |
| Neck Style | Tagless Crew |
| Sleeve Style | Raglan Sleeve |
| Finish | Ammonia Cure |
| Resistance | Flame |
| Hazard Risk Category | HRC1 |
| Arc Flash Rating | ATPV 6.4 Calories/cm2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.