Learn More
Bulwark™ FR Women's Classic Heavyweight Excel FR™ Flame Resistant Jean, Blue Denim
Shop All Red Kap Industries ProductsDescription
- Primary Closure: Zipper
- Wash Care: Industrial Laundry - Heavy Soil, Industrial Laundry - Light Soil, Home Wash
- Fabric: EXCEL FR™ Flame resistant, 14.75 oz. (500 g/m2)
- Blend: 100% Pre-Washed Cotton Denim
- Closure: One-piece waistband with button closure
- Pocket: Five jean-style pockets
- Features:
- One-piece waistband with button closure
- Five jean-style pockets
- Two-needle felled seam construction and soft pre-wash for comfort and look
- Classic fit is traditional, sits at the natural waist and has a slimmer leg opening that will fit over boots
- Product Collection: Excel FR
- Protection: ATPV Arc Rating: 21
- Country of Origin: Imported, Made in USA Fabric
- Fit Details: Sits at the waist. Regular in the seat and thigh. Slightly tapered leg.
- Gender: Female
- Materials: Cotton
- Brand: Bulwark™
Specifications
Specifications
| Size | 10 |
| Gender | Women's |
| Product Type | Flame-resistant Jeans |
| Material | Cotton |
| Color | Blue |
| Closure Type | Button |
| Hazard Risk Category | HRC2 |
| Certifications/Compliance | Meets NFPA 2112, UL |
| Size (English) Inseam | 32 in. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.